Assay Details
Target Gene Details
Entrez Gene ID: | 80830 |
Gene Name: | apolipoprotein L6 |
Gene Aliases: |
APOL-VI, APOLVI |
Location: |
Chr.22:35648317-35668409 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APOL6 | NM_030641.3 | NP_085144.1 | ||
| XM_011530392.2 | XP_011528694.1 | |||
| AK095881.1 | ||||
| AL834420.1 | CAH56356.1 | |||
| AX775805.1 | CAE11602.1 | |||
| AY014879.1 | AAK07724.1 | |||
| AY358210.1 | AAQ88577.1 | |||
| BC038950.2 | AAH38950.2 | |||
| BC047864.1 | AAH47864.1 | |||
| BU585026.1 | ||||
| CR456381.1 | CAG30267.1 |
More Information
Set Membership: |
|

Genomic Map