Assay Details
Target Gene Details
Entrez Gene ID: | 10201 |
Gene Name: | NME/NM23 nucleoside diphosphate kinase 6 |
Gene Aliases: |
IPIA-ALPHA, NDK 6, NM23-H6 |
Location: |
Chr.3:48289369-48302259 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NME6 | NM_001308426.1 | NP_001295355.1 | ||
| NM_001308427.1 | 1 | 72 | NP_001295356.1 | |
| NM_001308428.1 | 1 | 72 | NP_001295357.1 | |
| NM_001308430.1 | NP_001295359.1 | |||
| NM_001308431.1 | NP_001295360.1 | |||
| NM_001308433.1 | 1 | 72 | NP_001295362.1 | |
| NM_001308434.1 | NP_001295363.1 | |||
| NM_001308435.1 | NP_001295364.1 | |||
| NM_005793.4 | NP_005784.1 | |||
| XM_017005511.1 | XP_016861000.1 | |||
| XM_017005512.1 | XP_016861001.1 | |||
| XM_017005514.1 | 1 | 41 | XP_016861003.1 | |
| XM_017005515.1 | 1 | 41 | XP_016861004.1 | |
| XM_017005516.1 | XP_016861005.1 | |||
| XM_017005517.1 | XP_016861006.1 | |||
| AF051941.1 | AAC78463.1 | |||
| AK294809.1 | 1 | 28 | ||
| AK297364.1 | ||||
| AK308970.1 | ||||
| AK309090.1 | ||||
| AL699803.1 | 1 | 74 | ||
| BC001808.2 | AAH01808.1 | |||
| BC012828.2 | AAH12828.1 | |||
| BE866839.1 | ||||
| BG284082.1 | ||||
| BG530435.1 | ||||
| CA848473.1 | 1 | 51 | ||
| CD701581.1 | ||||
| CT002144.1 | ||||
| DA398008.1 | 1 | 28 | ||
| HY090220.1 | ||||
| U90449.1 | AAC69439.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834683 | Chr.3:48260014 - 48451855 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map