Assay Details
Target Gene Details
Entrez Gene ID: | 57447 |
Gene Name: | NDRG family member 2 |
Gene Aliases: |
SYLD |
Location: |
Chr.14:21016763-21070872 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDRG2 | NM_001282211.1 | NP_001269140.1 | ||
| NM_001282215.1 | NP_001269144.1 | |||
| NM_016250.2 | NP_057334.1 | |||
| NM_201535.1 | NP_963293.1 | |||
| NM_201536.1 | NP_963294.1 | |||
| NM_201537.1 | NP_963831.1 | |||
| XM_017021481.1 | XP_016876970.1 | |||
| XM_017021484.1 | XP_016876973.1 | |||
| XM_017021488.1 | XP_016876977.1 | |||
| XM_017021491.1 | XP_016876980.1 | |||
| XM_017021496.1 | XP_016876985.1 | |||
| AF304051.1 | AAL08624.1 | |||
| AK293514.1 | ||||
| BC010458.1 | AAH10458.1 | |||
| BC011240.1 | AAH11240.1 | |||
| BC093038.1 | AAH93038.1 | |||
| BG818977.1 | ||||
| CB153966.1 | 1 | 80 |
Target Gene Details
Entrez Gene ID: | 122664 |
Gene Name: | tubulin polymerization promoting protein family member 2 |
Gene Aliases: |
C14orf8, CT152, P18, p25beta |
Location: |
Chr.14:21024262-21036352 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TPPP2 | XM_017020966.1 | XP_016876455.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428293 | Chr.14:20747949 - 21063624 on Build GRCh38 | Gain+Loss |
|
| nsv832740 | Chr.14:20863901 - 21076893 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map