Assay Details
Target Gene Details
Entrez Gene ID: | 160492 |
Gene Name: | lamin tail domain containing 1 |
Gene Aliases: |
IFLTD1, PAS1C1 |
Location: |
Chr.12:25476082-25648604 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 18 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv437731 | Chr.12:25518825 - 25527233 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map