Assay Details
Target Gene Details
Entrez Gene ID: | 1655 |
Gene Name: | DEAD-box helicase 5 |
Gene Aliases: |
G17P1, HLR1, HUMP68, p68 |
Location: |
Chr.17:64498254-64507038 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDX5 | NM_001320595.1 | 14 | 2076 | NP_001307524.1 |
| NM_001320596.1 | 14 | 1965 | NP_001307525.1 | |
| NM_001320597.1 | 13 | 1816 | NP_001307526.1 | |
| NM_004396.4 | 13 | 2051 | NP_004387.1 | |
| AB209919.1 | 12 | 3038 | BAD93156.1 | |
| AB451257.1 | 13 | 1649 | ||
| AB451382.1 | 13 | 1649 | ||
| AK055995.1 | 2 | 1149 | ||
| AK297192.1 | 12 | 1581 | ||
| AK297753.1 | 14 | 1786 | ||
| AK297905.1 | 12 | 1609 | ||
| AK310318.1 | 13 | 1695 | ||
| BC016027.1 | 13 | 1790 | AAH16027.1 | |
| BT006943.1 | 13 | 1649 | AAP35589.1 | |
| BX571764.1 | 12 | 3056 | CAE11890.1 | |
| X15729.1 | 13 | 1816 | CAA33751.1 | |
| X52104.1 | 13 | 1824 | CAA36324.1 |
More Information
Set Membership: |
|

Genomic Map