Assay Details
Target Gene Details
Entrez Gene ID: | 55835 |
Gene Name: | centromere protein J |
Gene Aliases: |
BM032, CENP-J, CPAP, LAP, LIP1, MCPH6, SASS4, SCKL4, Sas-4 |
Location: |
Chr.13:24882274-24922889 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CENPJ | NM_018451.4 | 13 | 3598 | NP_060921.3 |
| NR_047594.1 | 14 | 3715 | ||
| NR_047595.1 | 12 | 3513 | ||
| AF139625.1 | 13 | 3613 | AAG21074.1 | |
| AF141337.1 | AAG49440.1 | |||
| AJ303006.1 | 5 | 574 | CAC80028.1 | |
| AK123551.1 | 2 | 991 | ||
| AK292406.1 | 13 | 3598 | ||
| AL833182.1 | 12 | 3460 | CAI46195.1 | |
| BC024209.2 | 6 | 632 | AAH24209.3 | |
| BC113110.1 | 13 | 3534 | AAI13111.1 | |
| BC113111.1 | 14 | 3687 | AAI13112.1 | |
| BC113662.1 | 13 | 3485 | ||
| BC113664.1 | 13 | 3485 |
Target Gene Details
Entrez Gene ID: | 56163 |
Gene Name: | ring finger protein 17 |
Gene Aliases: |
Mmip-2, SPATA23, TDRD4 |
Location: |
Chr.13:24747641-24888816 on Build GRCh38 |
Assay Gene Location: | Within Intron 43 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF17 | XM_011535156.2 | XP_011533458.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1041986 | Chr.13:24870991 - 24986494 on Build GRCh38 | Gain |
|
| nsv1053023 | Chr.13:24866993 - 24916871 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map