Assay Details
Target Gene Details
Entrez Gene ID: | 84986 |
Gene Name: | Rho GTPase activating protein 19 |
Gene Aliases: |
- |
Location: |
Chr.10:97222173-97292673 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGAP19 | NM_001204300.1 | 11 | 1790 | NP_001191229.1 |
| NM_001256423.1 | 12 | 1860 | NP_001243352.1 | |
| NM_032900.5 | 12 | 1877 | NP_116289.4 | |
| AK074122.1 | 7 | 957 | BAB84948.1 | |
| AK090447.1 | 13 | 1922 | BAC03428.1 | |
| AK093316.1 | 10 | 1508 | BAC04130.1 | |
| AK093441.1 | 12 | 1835 | BAC04165.1 | |
| AK303055.1 | ||||
| BC113888.1 | 11 | 1766 | ||
| BC114490.1 | 12 | 1853 | ||
| BX648189.1 | 12 | 1852 | ||
| BX648194.1 | 9 | 1320 | ||
| CR749412.1 | 12 | 1853 | CAH18254.1 | |
| DQ338460.1 | 12 | 1842 |
Target Gene Details
Entrez Gene ID: | 100533184 |
Gene Name: | ARHGAP19-SLIT1 readthrough (NMD candidate) |
Gene Aliases: |
- |
Location: |
Chr.10:97153042-97292673 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGAP19-SLIT1 | NR_037909.1 | |||
| AY336750.1 | AAR02412.1 |
More Information
Set Membership: |
|

Genomic Map