Assay Details
Target Gene Details
Entrez Gene ID: | 117144 |
Gene Name: | cation channel sperm associated 1 |
Gene Aliases: |
CATSPER, SPGF7 |
Location: |
Chr.11:66016752-66026518 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CATSPER1 | NM_053054.3 | 1 | 328 | NP_444282.3 |
| AF407333.1 | 1 | 190 | AAL14105.1 | |
| BC032950.2 | 1 | 328 | AAH32950.1 | |
| BC036522.2 | 1 | 290 | AAH36522.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832191 | Chr.11:65864858 - 66038649 on Build GRCh38 | Loss |
|
| nsv825954 | Chr.11:66025369 - 66035999 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map