Assay Details
Target Gene Details
Entrez Gene ID: | 27230 |
Gene Name: | stress-associated endoplasmic reticulum protein 1 |
Gene Aliases: |
RAMP4 |
Location: |
Chr.3:150541993-150603177 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SERP1 | NM_014445.3 | 3 | 738 | NP_055260.1 |
| XM_011512698.2 | 4 | 1403 | XP_011511000.1 | |
| AB022427.1 | 3 | 446 | BAA81895.1 | |
| AF136975.1 | 3 | 601 | AAG49436.1 | |
| AK090933.1 | 4 | 550 | ||
| AK093929.1 | 4 | 559 | ||
| AK098135.1 | 4 | 454 | ||
| AK125413.1 | 4 | 1504 | ||
| AL136807.1 | 3 | 547 | CAB66741.1 | |
| BC108314.1 | 3 | 586 | AAI08315.1 | |
| BC112364.1 | 3 | 545 | AAI12365.1 | |
| BC112365.1 | 3 | 545 | AAI12366.1 | |
| CR533472.1 | 3 | 232 | CAG38503.1 | |
| X75694.1 |
More Information
Set Membership: |
|

Genomic Map