Assay Details
Target Gene Details
Entrez Gene ID: | 79441 |
Gene Name: | HAUS augmin like complex subunit 3 |
Gene Aliases: |
C4orf15, IT1, dgt3 |
Location: |
Chr.4:2231836-2242164 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HAUS3 | NM_001303143.1 | 3 | 669 | NP_001290072.1 |
| NM_024511.6 | 2 | 398 | NP_078787.2 | |
| AF040964.1 | 1 | 142 | AAB97010.1 | |
| AK056134.1 | 2 | 886 | ||
| AK293948.1 | 2 | 886 | ||
| BC003648.2 | 3 | 669 | AAH03648.1 | |
| CR749640.1 | 2 | 367 | CAH18434.1 |
Target Gene Details
Entrez Gene ID: | 353497 |
Gene Name: | DNA polymerase nu |
Gene Aliases: |
POL4P |
Location: |
Chr.4:2071958-2242164 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLN | NM_181808.3 | NP_861524.2 | ||
| AK303673.1 | ||||
| DB098342.1 | ||||
| HY010559.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1009347 | Chr.4:2200935 - 2516063 on Build GRCh38 | Gain |
|
| esv2726835 | Chr.4:2019561 - 2959568 on Build GRCh38 | Deletion |
|
| nsv829838 | Chr.4:2155167 - 2349950 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map