Assay Details
Target Gene Details
Entrez Gene ID: | 64096 |
Gene Name: | GDNF family receptor alpha 4 |
Gene Aliases: |
- |
Location: |
Chr.20:3659292-3663399 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GFRA4 | NM_022139.3 | 6 | 1268 | NP_071422.1 |
| NM_145762.2 | 5 | 1358 | NP_665705.1 | |
| AF253318.1 | 6 | 1268 | AAG25925.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv520565 | Chr.20:3652593 - 3740054 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map