Assay Details
Target Gene Details
Entrez Gene ID: | 3788 |
Gene Name: | potassium voltage-gated channel modifier subfamily S member 2 |
Gene Aliases: |
KV9.2 |
Location: |
Chr.8:98427022-98430797 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNS2 | NM_020697.3 | 2 | 1616 | NP_065748.1 |
| AB032970.1 | 2 | 1421 | ||
| AK293096.1 | 1 | 1560 | ||
| BC027932.1 | 2 | 1589 | AAH27932.1 | |
| BC034778.1 | 2 | 1616 | AAH34778.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1034492 | Chr.8:98424113 - 98444288 on Build GRCh38 | Gain |
|
| nsv831406 | Chr.8:98278517 - 98469848 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map