Assay Details
Target Gene Details
Entrez Gene ID: | 3028 |
Gene Name: | hydroxysteroid 17-beta dehydrogenase 10 |
Gene Aliases: |
17b-HSD10, ABAD, CAMR, DUPXp11.22, ERAB, HADH2, HCD2, MHBD, MRPP2, MRX17, MRX31, MRXS10, SCHAD, SDR5C1 |
Location: |
Chr.X:53431258-53434376 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HSD17B10 | NM_001037811.2 | NP_001032900.1 | ||
| NM_004493.2 | NP_004484.1 | |||
| AF035555.2 | AAC15902.1 | |||
| AF069134.1 | AAC39900.1 | |||
| AU132847.1 | ||||
| AY092415.1 | AAM18189.1 | |||
| BC000372.2 | AAH00372.1 | |||
| BC008708.1 | AAH08708.1 | |||
| CR456723.1 | CAG33004.1 | |||
| KU178107.1 | ||||
| KU178108.1 | ||||
| U73514.1 | AAB68958.1 | |||
| U96132.1 | AAC51812.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2740175 | Chr.X:52862926 - 55652533 on Build GRCh38 | Deletion |
|
| nsv435891 | Chr.X:52861979 - 55653313 on Build GRCh38 | Deletion |
|
| nsv6908 | Chr.X:53412544 - 53446505 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map