Assay Details
Target Gene Details
Entrez Gene ID: | 9481 |
Gene Name: | solute carrier family 25 member 27 |
Gene Aliases: |
UCP4 |
Location: |
Chr.6:46652915-46678190 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC25A27 | NM_001204051.1 | 1 | 283 | NP_001190980.1 |
| NM_001204052.1 | 1 | 283 | NP_001190981.1 | |
| NM_004277.4 | 1 | 283 | NP_004268.3 | |
| XM_005249483.2 | 1 | 253 | XP_005249540.1 | |
| AF110532.1 | AAD16995.1 | |||
| AK291427.1 | 1 | 213 | ||
| AK295233.1 | 1 | 266 | ||
| AK302906.1 | 1 | 223 | ||
| AK315940.1 | 1 | 194 | ||
| AY358711.1 | AAQ89951.1 | |||
| BC033091.1 | 1 | 256 | AAH33091.1 | |
| BP310559.1 | ||||
| DA144842.1 | 1 | 257 | ||
| DC314392.1 | 1 | 283 | ||
| DR002070.1 | 1 | 205 |
More Information
Set Membership: |
|

Genomic Map