Assay Details
Target Gene Details
Entrez Gene ID: | 144132 |
Gene Name: | dynein heavy chain domain 1 |
Gene Aliases: |
C11orf47, CCDC35, DHCD1, DNHD1L |
Location: |
Chr.11:6497296-6572025 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNHD1 | NM_144666.2 | 3 | 233 | NP_653267.2 |
| NM_173589.3 | 2 | 336 | NP_775860.3 | |
| AK096944.1 | ||||
| BC111765.1 | 1 | 86 | AAI11766.1 | |
| BX648742.1 | 2 | 331 | ||
| DB099871.1 | 3 | 233 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv553372 | Chr.11:6489014 - 6529079 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map