Assay Details
Target Gene Details
Entrez Gene ID: | 6575 |
Gene Name: | solute carrier family 20 member 2 |
Gene Aliases: |
GLVR-2, GLVR2, IBGC1, IBGC3, MLVAR, PIT-2, PIT2, RAM1, Ram-1 |
Location: |
Chr.8:42416462-42542213 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC20A2 | NM_001257180.1 | 11 | 3169 | NP_001244109.1 |
| NM_001257181.1 | 11 | 2800 | NP_001244110.1 | |
| NM_006749.4 | 11 | 3132 | NP_006740.1 | |
| XM_005273613.3 | 11 | 2907 | XP_005273670.1 | |
| XM_005273615.3 | 12 | 2983 | XP_005273672.1 | |
| XM_006716390.3 | 10 | 2766 | XP_006716453.1 | |
| XM_006716391.3 | 12 | 6387 | XP_006716454.1 | |
| XM_017013748.1 | 11 | 3023 | XP_016869237.1 | |
| XM_017013749.1 | 10 | 6111 | XP_016869238.1 | |
| XM_017013750.1 | 12 | 3042 | XP_016869239.1 | |
| XM_017013751.1 | 12 | 2916 | XP_016869240.1 | |
| XM_017013752.1 | 12 | 3158 | XP_016869241.1 | |
| BC028600.2 | 11 | 2844 | AAH28600.1 | |
| L20852.1 | 10 | 2718 | AAA18018.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv396084 | Chr.8:42414652 - 42424106 on Build GRCh38 | Deletion |
|
| nsv831301 | Chr.8:42244080 - 42473666 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map