Assay Details
Target Gene Details
Entrez Gene ID: | 5284 |
Gene Name: | polymeric immunoglobulin receptor |
Gene Aliases: |
- |
Location: |
Chr.1:206928522-206946466 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIGR | NM_002644.3 | 11 | 2508 | NP_002635.2 |
| XM_011509629.1 | 11 | 2419 | XP_011507931.1 | |
| AF272149.2 | 11 | 2504 | AAN65630.1 | |
| AK225471.1 | ||||
| AY178835.1 | ||||
| BC110494.2 | AAI10495.1 | |||
| BC110495.2 | AAI10496.1 | |||
| CR749533.1 | 12 | 2581 | CAH18339.1 | |
| M24559.1 | 9 | 2111 | AAA36102.1 | |
| S62403.1 | 11 | 2480 | AAB20203.1 | |
| X73079.1 | 11 | 2504 | CAA51532.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1010486 | Chr.1:206876486 - 207031685 on Build GRCh38 | Gain |
|
| nsv468049 | Chr.1:206894495 - 206963993 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map