Assay Details
Target Gene Details
Entrez Gene ID: | 5532 |
Gene Name: | protein phosphatase 3 catalytic subunit beta |
Gene Aliases: |
CALNA2, CALNB, CNA2, PP2Bbeta |
Location: |
Chr.10:73436428-73496024 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP3CB | NM_001142353.2 | 14 | 1683 | NP_001135825.1 |
| NM_001142354.2 | 13 | 1653 | NP_001135826.1 | |
| NM_021132.3 | 14 | 1680 | NP_066955.1 | |
| XM_005269944.1 | 13 | 1650 | XP_005270001.1 | |
| XM_017016379.1 | 13 | 1585 | XP_016871868.1 | |
| AJ488506.1 | 13 | 1590 | CAD32694.1 | |
| AK300701.1 | 6 | 476 | ||
| BC028049.1 | 14 | 1671 | AAH28049.1 | |
| M29551.1 | 14 | 1661 | AAA35706.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831911 | Chr.10:73313880 - 73483446 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map