Assay Details
Target Gene Details
Entrez Gene ID: | 130916 |
Gene Name: | mitochondrial transcription termination factor 4 |
Gene Aliases: |
MTERFD2 |
Location: |
Chr.2:241072168-241103367 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTERF4 | NR_028049.1 | 7 | 3244 | |
| NR_028050.1 | 5 | 1436 | ||
| NR_028051.1 | 4 | 1251 | ||
| XM_006712294.1 | 5 | 1705 | XP_006712357.1 | |
| XM_006712295.3 | 5 | 1861 | XP_006712358.1 | |
| XM_011510634.2 | 7 | 3547 | XP_011508936.1 | |
| XM_017003379.1 | 7 | 4175 | XP_016858868.1 | |
| XM_017003380.1 | XP_016858869.1 | |||
| XM_017003381.1 | 8 | 4011 | XP_016858870.1 | |
| XM_017003382.1 | 8 | 3524 | XP_016858871.1 | |
| XM_017003383.1 | 8 | 4297 | XP_016858872.1 | |
| XM_017003384.1 | 8 | 4272 | XP_016858873.1 | |
| XM_017003385.1 | XP_016858874.1 | |||
| XM_017003386.1 | 7 | 4646 | XP_016858875.1 | |
| XM_017003387.1 | 6 | 4198 | XP_016858876.1 | |
| XM_017003389.1 | 6 | 2958 | XP_016858878.1 | |
| XM_017003390.1 | 5 | 3431 | XP_016858879.1 | |
| XM_017003391.1 | 5 | 4451 | XP_016858880.1 | |
| AK025374.1 | 1 | 496 | ||
| AK055474.1 | 4 | 2641 | ||
| AK093695.1 | 5 | 1391 | ||
| AK124444.1 | ||||
| BX648184.1 | 7 | 3210 |
Target Gene Details
Entrez Gene ID: | 25992 |
Gene Name: | sushi, nidogen and EGF like domains 1 |
Gene Aliases: |
SST3, Snep |
Location: |
Chr.2:240997409-241095568 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 37 - Intron 37 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNED1 | NM_001080437.1 | NP_001073906.1 | ||
| XM_011510931.2 | XP_011509233.1 | |||
| XM_011510932.2 | XP_011509234.1 | |||
| AF439717.1 | AAQ04558.1 | |||
| AK074062.1 | BAB84888.1 | |||
| AK074075.1 | BAB84901.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3575345 | Chr.2:241086207 - 241153068 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map