Assay Details
Target Gene Details
Entrez Gene ID: | 6590 |
Gene Name: | secretory leukocyte peptidase inhibitor |
Gene Aliases: |
ALK1, ALP, BLPI, HUSI, HUSI-I, MPI, WAP4, WFDC4 |
Location: |
Chr.20:45252239-45254564 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLPI | NM_003064.3 | NP_003055.1 | ||
| AF114471.1 | AAD19661.1 | |||
| AK312192.1 | ||||
| AX772818.1 | CAE06258.1 | |||
| BC020708.1 | AAH20708.1 | |||
| X04470.1 | CAA28158.1 | |||
| X04503.1 | CAA28188.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1056102 | Chr.20:44673341 - 45636248 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map