Assay Details
Target Gene Details
Entrez Gene ID: | 155185 |
Gene Name: | archaelysin family metallopeptidase 1 |
Gene Aliases: |
- |
Location: |
Chr.7:2679522-2767486 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AMZ1 | NM_001321766.1 | NP_001308695.1 | ||
| XM_011515151.2 | XP_011513453.1 | |||
| XM_017011774.1 | XP_016867263.1 | |||
| AK308900.1 |
Target Gene Details
Entrez Gene ID: | 2768 |
Gene Name: | G protein subunit alpha 12 |
Gene Aliases: |
NNX3, RMP, gep |
Location: |
Chr.7:2728105-2870656 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GNA12 | NM_001282440.1 | 3 | 1449 | NP_001269369.1 |
| NM_001282441.1 | 5 | 1577 | NP_001269370.1 | |
| NM_001293092.1 | 3 | 1538 | NP_001280021.1 | |
| NM_007353.2 | 4 | 1589 | NP_031379.2 | |
| XM_011515288.2 | 4 | 1149 | XP_011513590.1 | |
| XM_017012008.1 | 4 | 1472 | XP_016867497.1 | |
| AK025582.1 | 1 | 579 | ||
| AK025894.1 | 1 | 64 | ||
| AK122830.1 | 4 | 1361 | ||
| AK127842.1 | 5 | 1577 | ||
| AK128701.1 | 3 | 1449 | BAC87579.1 | |
| AK295830.1 | 3 | 1229 | ||
| BC087537.1 | 4 | 1602 | AAH87537.1 | |
| BC111464.1 | 4 | 1593 | AAI11465.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3611852 | Chr.7:2719771 - 2731488 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map