Assay Details
Target Gene Details
Entrez Gene ID: | 81493 |
Gene Name: | syncoilin, intermediate filament protein |
Gene Aliases: |
SYNC1, SYNCOILIN |
Location: |
Chr.1:32679906-32702760 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SYNC | NM_001161708.1 | 2 | 1165 | NP_001155180.1 |
| NM_030786.2 | 2 | 1165 | NP_110413.2 | |
| AA376455.1 | ||||
| AK024707.1 | 1 | 240 | BAB14970.1 | |
| AK297958.1 | 2 | 1161 | ||
| AK302275.1 | 2 | 1069 | ||
| BC119700.1 | 1 | 135 | ||
| BC119701.1 | 1 | 135 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509158 | Chr.1:32691713 - 32783820 on Build GRCh38 | Insertion |
|
| nsv519188 | Chr.1:32619643 - 32780839 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map