Assay Details
Target Gene Details
Entrez Gene ID: | 2134 |
Gene Name: | exostosin like glycosyltransferase 1 |
Gene Aliases: |
EXTL |
Location: |
Chr.1:26021749-26036464 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EXTL1 | NM_004455.2 | 11 | 3183 | NP_004446.2 |
| XM_005245779.3 | 10 | 3106 | XP_005245836.1 | |
| BC065528.1 | 11 | 3029 | AAH65528.1 | |
| U67191.1 | 11 | 3182 | AAC51141.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834513 | Chr.1:25922980 - 26087385 on Build GRCh38 | Gain |
|
| nsv950338 | Chr.1:26031610 - 26047909 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map