Assay Details
Target Gene Details
Entrez Gene ID: | 783 |
Gene Name: | calcium voltage-gated channel auxiliary subunit beta 2 |
Gene Aliases: |
CACNLB2, CAVB2, MYSB |
Location: |
Chr.10:18140560-18545881 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
Target Gene Details
Entrez Gene ID: | 221078 |
Gene Name: | NOP2/Sun RNA methyltransferase family member 6 |
Gene Aliases: |
4933414E04Rik, ARL5B-AS1, NOPD1 |
Location: |
Chr.10:18512582-18659264 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NSUN6 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1042073 | Chr.10:18517971 - 18549374 on Build GRCh38 | Gain |
|
| nsv550042 | Chr.10:18394338 - 18705654 on Build GRCh38 | Gain |
|
| esv34545 | Chr.10:18532256 - 18619365 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map