Assay Details
Target Gene Details
Entrez Gene ID: | 65987 |
Gene Name: | potassium channel tetramerization domain containing 14 |
Gene Aliases: |
- |
Location: |
Chr.11:78015715-78046191 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCTD14 | NM_001282406.1 | 3 | 1315 | NP_001269335.1 |
| NM_023930.3 | 2 | 1204 | NP_076419.2 | |
| BC001062.2 | 2 | 1170 | AAH01062.1 | |
| BC001929.1 | 2 | 1170 | AAH01929.1 | |
| BU631809.1 | 1 | 485 |
Target Gene Details
Entrez Gene ID: | 100532726 |
Gene Name: | NDUFC2-KCTD14 readthrough |
Gene Aliases: |
- |
Location: |
Chr.11:78015715-78080219 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFC2-KCTD14 | NM_001203260.1 | 4 | 1969 | NP_001190189.1 |
| NM_001203261.1 | 3 | 1874 | NP_001190190.1 | |
| NM_001203262.1 | 3 | 1825 | NP_001190191.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv520740 | Chr.11:77842592 - 78286890 on Build GRCh38 | Gain |
|
| nsv516638 | Chr.11:78013628 - 78052743 on Build GRCh38 | Loss |
|
| esv3626891 | Chr.11:78014218 - 78019545 on Build GRCh38 | Loss |
|
| nsv1050891 | Chr.11:77705600 - 78076956 on Build GRCh38 | Gain |
|
| esv2660024 | Chr.11:78014144 - 78019622 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map