Assay Details
Target Gene Details
Entrez Gene ID: | 29890 |
Gene Name: | RNA binding motif protein 15B |
Gene Aliases: |
HUMAGCGB, OTT3 |
Location: |
Chr.3:51391268-51397908 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RBM15B | NM_013286.4 | 1 | 4311 | NP_037418.3 |
| AL831838.1 | 1 | 3717 | CAD38547.2 | |
| BG674080.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv590277 | Chr.3:51370620 - 51466575 on Build GRCh38 | Loss |
|
| nsv590276 | Chr.3:51358140 - 51431657 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map