Assay Details
Target Gene Details
Entrez Gene ID: | 84867 |
Gene Name: | protein tyrosine phosphatase, non-receptor type 5 |
Gene Aliases: |
PTPSTEP, STEP, STEP61 |
Location: |
Chr.11:18727928-18792721 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1047371 | Chr.11:18709926 - 18741490 on Build GRCh38 | Gain |
|
| nsv553633 | Chr.11:18290154 - 19036904 on Build GRCh38 | Gain |
|
| nsv525192 | Chr.11:18701244 - 18771813 on Build GRCh38 | Loss |
|
| nsv525452 | Chr.11:18714400 - 18780956 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map