Assay Details
Target Gene Details
Entrez Gene ID: | 11163 |
Gene Name: | nudix hydrolase 4 |
Gene Aliases: |
DIPP2, DIPP2alpha, DIPP2beta, HDCMB47P |
Location: |
Chr.12:93377923-93403248 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUDT4 | NM_001301022.1 | 5 | 3863 | NP_001287951.1 |
| NM_001301023.1 | 5 | 3860 | NP_001287952.1 | |
| NM_001301024.1 | 5 | 3763 | NP_001287953.1 | |
| NM_019094.5 | 5 | 4118 | NP_061967.3 | |
| NM_199040.3 | 5 | 4121 | NP_950241.1 | |
| XM_011537794.2 | 5 | 3661 | XP_011536096.1 | |
| XM_011537795.2 | 6 | 5673 | XP_011536097.1 | |
| XM_011537796.2 | 6 | 3802 | XP_011536098.1 | |
| AF191651.1 | 5 | 3709 | AAF68857.1 | |
| AF191652.2 | 5 | 3712 | AAF68858.2 | |
| AF191655.1 | 1 | 952 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1044304 | Chr.12:93219056 - 93419239 on Build GRCh38 | Gain |
|
| nsv952465 | Chr.12:93398225 - 93408024 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map