Assay Details
Target Gene Details
Entrez Gene ID: | 55152 |
Gene Name: | DALR anticodon binding domain containing 3 |
Gene Aliases: |
- |
Location: |
Chr.3:49015399-49021386 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DALRD3 | NM_001009996.2 | NP_001009996.1 | ||
| NM_001276405.1 | NP_001263334.1 | |||
| NM_018114.5 | NP_060584.3 | |||
| XM_005265269.3 | XP_005265326.1 | |||
| XM_006713219.2 | XP_006713282.1 | |||
| XM_011533891.1 | XP_011532193.1 | |||
| XM_011533892.1 | XP_011532194.1 | |||
| XM_011533893.1 | XP_011532195.1 | |||
| XM_017006723.1 | XP_016862212.1 | |||
| AK001358.1 | BAA91647.1 | |||
| AK093054.1 | ||||
| AK093204.1 | BAC04095.1 | |||
| AK093294.1 | BAC04123.1 | |||
| AK301352.1 | ||||
| AK308273.1 | ||||
| BC032440.1 | AAH32440.2 | |||
| BC047683.1 | AAH47683.1 | |||
| BC054493.1 | AAH54493.1 | |||
| BX648719.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv954488 | Chr.3:49002068 - 49019667 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map