Assay Details
Target Gene Details
Entrez Gene ID: | 314 |
Gene Name: | amine oxidase, copper containing 2 |
Gene Aliases: |
DAO2, RAO |
Location: |
Chr.17:42844592-42850707 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AOC2 | NM_001158.3 | 1 | 1409 | NP_001149.2 |
| NM_009590.2 | 1 | 1409 | NP_033720.2 | |
| AF081363.1 | 1 | 1400 | AAD39345.1 | |
| BC142641.1 | 1 | 1401 | ||
| D88213.1 | 1 | 1400 | BAA19001.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv483009 | Chr.17:42829470 - 43006887 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map