Assay Details
Target Gene Details
Entrez Gene ID: | 9852 |
Gene Name: | EPM2A interacting protein 1 |
Gene Aliases: |
- |
Location: |
Chr.3:36985866-36993304 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EPM2AIP1 | NM_014805.3 | 1 | 3198 | NP_055620.1 |
| AB018309.1 | 1 | 3087 | ||
| AK002069.1 | 1 | 2011 | ||
| AY178832.1 | 1 | 3198 | AAO21376.1 | |
| BC064696.1 | 1 | 3062 | AAH64696.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834662 | Chr.3:36830593 - 37033470 on Build GRCh38 | Loss |
|
| nsv954463 | Chr.3:36984810 - 36995509 on Build GRCh38 | Duplication |
|
| nsv834663 | Chr.3:36966258 - 37148914 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map