Assay Details
Target Gene Details
Entrez Gene ID: | 284361 |
Gene Name: | ER membrane protein complex subunit 10 |
Gene Aliases: |
C19orf63, HSM1, HSS1 |
Location: |
Chr.19:50476477-50505905 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EMC10 | NM_175063.5 | 8 | 1612 | NP_778233.4 |
| NM_206538.3 | 7 | 1525 | NP_996261.1 | |
| XM_006723163.3 | XP_006723226.1 | |||
| XM_011526818.2 | XP_011525120.1 | |||
| AL512761.1 | 4 | 1128 | CAH10658.1 | |
| AY194293.2 | 12 | 2570 | ||
| AY358710.1 | AAQ89073.1 | |||
| AY761095.1 | 8 | 1590 | AAV30543.1 | |
| AY761097.1 | 7 | 1503 | AAV30545.1 | |
| BC032948.2 | 7 | 1466 | AAH32948.1 | |
| BC035001.2 | 7 | 1464 | AAH35001.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509750 | Chr.19:50443078 - 50540357 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map