Assay Details
Target Gene Details
Entrez Gene ID: | 2023 |
Gene Name: | enolase 1 |
Gene Aliases: |
ENO1L1, HEL-S-17, MPB1, NNE, PPH |
Location: |
Chr.1:8861000-8879092 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ENO1 | NM_001201483.1 | NP_001188412.1 | ||
| NM_001428.3 | NP_001419.1 | |||
| XM_006710433.2 | XP_006710496.1 | |||
| AF035286.1 | AAB88178.1 | |||
| AK098420.1 | ||||
| AK222517.1 | BAD96237.1 | |||
| AK223192.1 | BAD96912.1 | |||
| AK309177.1 | ||||
| AK315417.1 | ||||
| AL833741.1 | CAH56247.1 | |||
| BC001810.2 | AAH01810.1 | |||
| BC004325.1 | AAH04325.1 | |||
| BC004458.1 | AAH04458.1 | |||
| BC009912.2 | AAH09912.1 | |||
| BC011130.1 | AAH11130.1 | |||
| BC015641.2 | AAH15641.1 | |||
| BC021166.2 | AAH21166.2 | |||
| BC022545.1 | AAH22545.1 | |||
| BC027725.2 | AAH27725.1 | |||
| BC050642.2 | AAH50642.1 | |||
| BT007163.1 | AAP35827.1 | |||
| BX537400.1 | CAD97642.1 | |||
| GQ472226.1 | ||||
| GU170215.1 | ||||
| M14328.1 | AAA52387.1 | |||
| M55914.1 | AAA35698.1 | |||
| X84907.1 | CAA59331.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv545345 | Chr.1:8831066 - 9035551 on Build GRCh38 | Gain |
|
| nsv1001865 | Chr.1:8843872 - 8873183 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map