Assay Details
Target Gene Details
Entrez Gene ID: | 80185 |
Gene Name: | TELO2 interacting protein 2 |
Gene Aliases: |
C8orf41, MRT39 |
Location: |
Chr.8:33498509-33513601 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTI2 | NM_001102401.2 | 5 | 1267 | NP_001095871.1 |
| NM_001265581.1 | 5 | 1389 | NP_001252510.1 | |
| NM_025115.3 | 4 | 1636 | NP_079391.2 | |
| XM_011544665.2 | 4 | 1590 | XP_011542967.1 | |
| AK026916.1 | 4 | 1489 | BAB15590.1 | |
| BC007387.2 | 5 | 1192 | AAH07387.1 | |
| BC066935.1 | 5 | 1220 | AAH66935.1 | |
| BX648410.1 | 5 | 1303 |

Genomic Map