Assay Details
Target Gene Details
Entrez Gene ID: | 117245 |
Gene Name: | HRAS like suppressor family member 5 |
Gene Aliases: |
HRLP5, HRSL5, RLP1, iNAT |
Location: |
Chr.11:63461404-63491208 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HRASLS5 | NM_001146728.1 | 6 | 972 | NP_001140200.1 |
| NM_001146729.1 | 6 | 992 | NP_001140201.1 | |
| NM_054108.3 | 6 | 1022 | NP_473449.1 | |
| XM_011544751.2 | 6 | 927 | XP_011543053.1 | |
| XM_011544752.2 | 3 | 627 | XP_011543054.1 | |
| XM_011544753.1 | 7 | 846 | XP_011543055.1 | |
| XM_017017172.1 | 7 | 834 | XP_016872661.1 | |
| AB298804.1 | ||||
| AJ416558.1 | 6 | 1009 | CAC94942.1 | |
| AK055392.1 | 4 | 508 | ||
| AK302312.1 | 6 | 933 | ||
| BC034222.1 | 6 | 951 | AAH34222.1 |
More Information
Set Membership: |
|

Genomic Map