Assay Details
Target Gene Details
Entrez Gene ID: | 81831 |
Gene Name: | neuropilin and tolloid like 2 |
Gene Aliases: |
BTCL2, NEOT2 |
Location: |
Chr.16:47081520-47144025 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NETO2 | NM_001201477.1 | NP_001188406.1 | ||
| NM_018092.4 | NP_060562.3 | |||
| XM_006721289.1 | XP_006721352.1 | |||
| XM_006721291.1 | XP_006721354.1 | |||
| XM_017023740.1 | XP_016879229.1 | |||
| AK027630.1 | BAB55247.1 | |||
| AK074937.1 | BAC11303.1 | |||
| AK297408.1 | ||||
| AL834404.1 | CAD39066.1 | |||
| AY358718.1 | AAQ89080.1 | |||
| BC012381.1 | AAH12381.1 | |||
| BC108727.1 | AAI08728.1 | |||
| BX538063.1 | CAD97994.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1066115 | Chr.16:46429858 - 47284504 on Build GRCh38 | Gain |
|
| esv2659121 | Chr.16:47084567 - 47087932 on Build GRCh38 | Deletion |
|
| esv3638547 | Chr.16:47084579 - 47088036 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map