Assay Details
Target Gene Details
Entrez Gene ID: | 581 |
Gene Name: | BCL2 associated X, apoptosis regulator |
Gene Aliases: |
BCL2L4 |
Location: |
Chr.19:48954825-48961798 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BAX | NM_001291428.1 | 4 | 374 | NP_001278357.1 |
| NM_001291429.1 | 3 | 322 | NP_001278358.1 | |
| NM_001291430.1 | 2 | 175 | NP_001278359.1 | |
| NM_001291431.1 | 2 | 175 | NP_001278360.1 | |
| NM_004324.3 | 4 | 374 | NP_004315.1 | |
| NM_138761.3 | 4 | 374 | NP_620116.1 | |
| NM_138763.3 | 3 | 227 | NP_620118.1 | |
| NM_138764.4 | 4 | 374 | NP_620119.2 | |
| NR_027882.1 | 4 | 374 | ||
| XM_017027077.1 | 4 | 383 | XP_016882566.1 | |
| AF007826.1 | 4 | 305 | AAD22706.1 | |
| AF247393.1 | 4 | 305 | AAF71267.1 | |
| AF250190.1 | 2 | 106 | AAF82094.1 | |
| AJ417988.1 | 3 | 284 | CAD10744.1 | |
| AJ586909.1 | 2 | 93 | CAE52909.1 | |
| AJ586910.1 | 3 | 240 | CAE52910.1 | |
| AK291076.1 | 4 | 354 | ||
| BC014175.2 | 4 | 357 | AAH14175.1 | |
| BE396495.1 | 4 | 374 | ||
| BM706954.1 | 2 | 187 | ||
| CN281003.1 | 4 | 327 | ||
| HQ889830.1 | 2 | 106 | ||
| JX524562.1 | 3 | 254 | ||
| KF830846.1 | 3 | 254 | ||
| KF830847.1 | 3 | 254 | ||
| KF830848.1 | 3 | 254 | ||
| L22473.1 | 4 | 305 | AAA03619.1 | |
| L22474.1 | 4 | 305 | AAA03620.1 | |
| T29044.1 | ||||
| U19599.1 | 3 | 158 | AAC50142.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss |
|
| dgv3618n100 | Chr.19:48948950 - 49021211 on Build GRCh38 | Gain |
|
| nsv833857 | Chr.19:48914476 - 49146320 on Build GRCh38 | Loss |
|
| nsv833856 | Chr.19:48847866 - 48986483 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map