Assay Details
Target Gene Details
Entrez Gene ID: | 26354 |
Gene Name: | G protein nucleolar 3 |
Gene Aliases: |
C77032, E2IG3, NNP47, NS |
Location: |
Chr.3:52685920-52694497 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GNL3 | NM_014366.4 | NP_055181.3 | ||
| NM_206825.1 | NP_996561.1 | |||
| NM_206826.1 | NP_996562.1 | |||
| AF191018.1 | AAF09482.1 | |||
| AK027514.1 | BAB55168.1 | |||
| AK027516.1 | BAB55169.1 | |||
| AK315484.1 | ||||
| AY825265.1 | AAV74413.1 | |||
| BC001024.2 | AAH01024.1 | |||
| CF272475.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3596161 | Chr.3:52690398 - 52692540 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map