Assay Details
Target Gene Details
Entrez Gene ID: | 10063 |
Gene Name: | COX17, cytochrome c oxidase copper chaperone |
Gene Aliases: |
- |
Location: |
Chr.3:119659975-119677441 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COX17 |
Target Gene Details
Entrez Gene ID: | 64091 |
Gene Name: | popeye domain containing 2 |
Gene Aliases: |
POP2 |
Location: |
Chr.3:119642052-119660590 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POPDC2 | NM_001308333.1 | 1 | 222 | NP_001295262.1 |
| NM_022135.3 | 1 | 222 | NP_071418.2 | |
| XM_005247698.3 | 1 | 222 | XP_005247755.1 | |
| AF204173.1 | 1 | 205 | AAG23406.1 | |
| AK226022.1 | 2 | 287 | ||
| AK314819.1 | 1 | 223 | ||
| AK377174.1 | ||||
| AK377176.1 | ||||
| AK377177.1 | ||||
| AK377178.1 | ||||
| AK377179.1 | ||||
| AK377180.1 | ||||
| AK377181.1 | ||||
| AK377182.1 | ||||
| AK377183.1 | ||||
| AK377184.1 | ||||
| BC026911.2 | AAH26911.1 | |||
| BC044929.1 | 1 | 222 | AAH44929.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1014394 | Chr.3:119641659 - 119693428 on Build GRCh38 | Loss |
|
| esv3568977 | Chr.3:119657752 - 119694492 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map