Assay Details
Target Gene Details
Entrez Gene ID: | 133308 |
Gene Name: | solute carrier family 9 member B2 |
Gene Aliases: |
NHA2, NHE10, NHEDC2 |
Location: |
Chr.4:103018033-103077371 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC9B2 | NM_001300754.1 | NP_001287683.1 | ||
| NM_001300756.1 | NP_001287685.1 | |||
| NM_178833.5 | NP_849155.2 | |||
| XM_006714085.2 | XP_006714148.1 | |||
| XM_006714086.3 | XP_006714149.1 | |||
| AA233228.1 | ||||
| AK172823.1 | BAD18790.1 | |||
| AK297010.1 | ||||
| AK299888.1 | ||||
| BC009732.1 | AAH09732.1 | |||
| BC047447.1 | AAH47447.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4445 | Chr.4:103003411 - 103034963 on Build GRCh38 | Insertion |
|
| nsv594978 | Chr.4:102886563 - 103032232 on Build GRCh38 | Loss |
|
| esv2759270 | Chr.4:102859548 - 103136164 on Build GRCh38 | Gain |
|
| nsv830021 | Chr.4:103013297 - 103179431 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map