Assay Details
Target Gene Details
Entrez Gene ID: | 121260 |
Gene Name: | solute carrier family 15 member 4 |
Gene Aliases: |
FP12591, PHT1, PTR4 |
Location: |
Chr.12:128793194-128823996 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC15A4 | NM_145648.3 | NP_663623.1 | ||
| XM_011537895.1 | XP_011536197.1 | |||
| AF461893.1 | AAQ04807.1 | |||
| AK095717.1 | ||||
| AK097154.1 | 1 | 913 | ||
| AY038999.1 | AAK72099.1 | |||
| AY050629.1 | AAK95565.1 | |||
| BC028394.2 | AAH28394.2 | |||
| BC146803.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1051433 | Chr.12:128679360 - 128888555 on Build GRCh38 | Gain |
|
| dgv1576n100 | Chr.12:128763841 - 128795043 on Build GRCh38 | Gain |
|
| esv3631151 | Chr.12:128793327 - 128853915 on Build GRCh38 | Gain |
|
| esv2759920 | Chr.12:128587760 - 128896765 on Build GRCh38 | Loss |
|
| nsv560650 | Chr.12:128744185 - 128799166 on Build GRCh38 | Gain |
|
| nsv1040831 | Chr.12:128749443 - 129516029 on Build GRCh38 | Gain |
|
| dgv527e212 | Chr.12:128794319 - 128806710 on Build GRCh38 | Loss |
|
| esv3581993 | Chr.12:128679608 - 128885566 on Build GRCh38 | Gain |
|
| nsv832549 | Chr.12:128739165 - 128936554 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs147786891] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map