Assay Details
Target Gene Details
Entrez Gene ID: | 9016 |
Gene Name: | solute carrier family 25 member 14 |
Gene Aliases: |
BMCP1, UCP5 |
Location: |
Chr.X:130339888-130373361 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC25A14 | NM_001282195.1 | 2 | 148 | NP_001269124.1 |
| NM_001282196.1 | 2 | 148 | NP_001269125.1 | |
| NM_001282197.1 | 2 | 148 | NP_001269126.1 | |
| NM_001282198.1 | NP_001269127.1 | |||
| NR_104107.1 | ||||
| XM_005262489.4 | XP_005262546.1 | |||
| XM_011531402.2 | 2 | 123 | XP_011529704.1 | |
| XM_011531403.1 | XP_011529705.1 | |||
| XM_017029936.1 | XP_016885425.1 | |||
| XM_017029937.1 | 2 | 123 | XP_016885426.1 | |
| XM_017029938.1 | 2 | 145 | XP_016885427.1 | |
| XM_017029939.1 | 2 | 123 | XP_016885428.1 | |
| XM_017029940.1 | XP_016885429.1 | |||
| XM_017029941.1 | 2 | 123 | XP_016885430.1 | |
| AF078544.1 | 1 | 144 | AAD04346.1 | |
| AK296155.1 | ||||
| AK297499.1 | 2 | 82 | ||
| AK304649.1 | ||||
| AK309433.1 | 2 | 82 | ||
| AL520510.3 | ||||
| BC119666.2 | ||||
| BC119667.1 | ||||
| DB485834.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528290 | Chr.X:129635682 - 130429852 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map