Assay Details
Target Gene Details
Entrez Gene ID: | 9154 |
Gene Name: | solute carrier family 28 member 1 |
Gene Aliases: |
CNT1, HCNT1 |
Location: |
Chr.15:84884351-84975649 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC28A1 | NM_001287761.1 | NP_001274690.1 | ||
| NM_001287762.1 | NP_001274691.1 | |||
| NM_001321721.1 | NP_001308650.1 | |||
| NM_001321722.1 | NP_001308651.1 | |||
| NM_004213.4 | NP_004204.3 | |||
| NM_201651.2 | NP_964014.1 | |||
| XM_011522203.2 | XP_011520505.1 | |||
| XM_011522204.2 | 1 | 339 | XP_011520506.1 | |
| XM_011522205.2 | 1 | 339 | XP_011520507.1 | |
| XM_011522206.2 | 1 | 339 | XP_011520508.1 | |
| XM_011522208.2 | 1 | 347 | XP_011520510.1 | |
| XM_011522209.2 | XP_011520511.1 | |||
| XM_011522210.2 | XP_011520512.1 | |||
| XM_011522211.2 | 1 | 344 | XP_011520513.1 | |
| XM_011522212.1 | XP_011520514.1 | |||
| XM_011522214.2 | XP_011520516.1 | |||
| XM_011522215.2 | XP_011520517.1 | |||
| XM_011522217.1 | XP_011520519.1 | |||
| XM_011522218.2 | XP_011520520.1 | |||
| XM_017022723.1 | XP_016878212.1 | |||
| AF309632.1 | AAL09447.1 | |||
| AK291997.1 | ||||
| AK296074.1 | ||||
| AK296075.1 | ||||
| AK296119.1 | ||||
| AK298369.1 | ||||
| AK309571.1 | ||||
| AK309578.1 | ||||
| AK311387.1 | ||||
| BC029788.1 | AAH29788.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524050 | Chr.15:84864333 - 84935498 on Build GRCh38 | Loss |
|
| esv3637100 | Chr.15:84883478 - 84885266 on Build GRCh38 | Loss |
|
| esv2673757 | Chr.15:84883448 - 84885252 on Build GRCh38 | Deletion |
|
| esv3637099 | Chr.15:84858948 - 84903602 on Build GRCh38 | Gain |
|
| dgv322n27 | Chr.15:84869224 - 84919085 on Build GRCh38 | Gain |
|
| esv2672879 | Chr.15:84183834 - 85188520 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map