Assay Details
Target Gene Details
Entrez Gene ID: | 84899 |
Gene Name: | transmembrane and tetratricopeptide repeat containing 4 |
Gene Aliases: |
- |
Location: |
Chr.13:100603836-100675025 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMTC4 | NM_001079669.2 | 18 | 3307 | NP_001073137.1 |
| NM_001286453.1 | 16 | 2974 | NP_001273382.1 | |
| NM_032813.3 | 19 | 3517 | NP_116202.2 | |
| XM_011521121.2 | 20 | 3669 | XP_011519423.1 | |
| XM_011521122.2 | 20 | 3585 | XP_011519424.1 | |
| XM_011521123.2 | 20 | 3746 | XP_011519425.1 | |
| XM_011521124.2 | 19 | 3374 | XP_011519426.1 | |
| XM_017020796.1 | 19 | 3572 | XP_016876285.1 | |
| XM_017020797.1 | 18 | 3311 | XP_016876286.1 | |
| XM_017020798.1 | 18 | 3407 | XP_016876287.1 | |
| XM_017020799.1 | 17 | 3092 | XP_016876288.1 | |
| AF339792.1 | 16 | 2889 | ||
| AK027530.1 | 13 | 2400 | BAB55179.1 | |
| AK056409.1 | 19 | 3376 | ||
| AK124599.1 | 1 | 3978 | ||
| AW513848.1 | 1 | 341 | ||
| BX647956.1 | 18 | 3198 | ||
| BX648008.1 | 18 | 3151 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1037414 | Chr.13:100521000 - 100622872 on Build GRCh38 | Loss |
|

Genomic Map