Assay Details
Target Gene Details
Entrez Gene ID: | 10352 |
Gene Name: | tryptophanyl tRNA synthetase 2, mitochondrial |
Gene Aliases: |
TrpRS |
Location: |
Chr.1:119031216-119140701 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WARS2 | NM_015836.3 | 6 | 1008 | NP_056651.1 |
| NM_201263.2 | 6 | 1037 | NP_957715.1 | |
| XM_005270350.3 | 6 | 1094 | XP_005270407.1 | |
| XM_011540494.2 | 7 | 1077 | XP_011538796.1 | |
| XM_011540495.2 | 5 | 779 | XP_011538797.1 | |
| XM_017000038.1 | 5 | 943 | XP_016855527.1 | |
| XM_017000039.1 | 8 | 1263 | XP_016855528.1 | |
| XM_017000040.1 | 6 | 830 | XP_016855529.1 | |
| XM_017000041.1 | 5 | 920 | XP_016855530.1 | |
| XM_017000042.1 | 5 | 914 | XP_016855531.1 | |
| AJ242739.1 | 6 | 989 | CAB63107.1 | |
| AK223197.1 | 6 | 986 | BAD96917.1 | |
| AK296799.1 | 5 | 903 | ||
| AK299547.1 | 6 | 947 | ||
| AK300295.1 | 6 | 836 | ||
| AK313740.1 | 6 | 1006 | ||
| BC044575.1 | 6 | 991 | AAH44575.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv997707 | Chr.1:118656945 - 119053326 on Build GRCh38 | Gain |
|
| nsv524681 | Chr.1:118890895 - 121570001 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map