Assay Details
Target Gene Details
Entrez Gene ID: | 100529144 |
Gene Name: | CORO7-PAM16 readthrough |
Gene Aliases: |
- |
Location: |
Chr.16:4340251-4416961 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 28 - Exon 28 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CORO7-PAM16 | NM_001201479.1 | NP_001188408.1 | ||
| AL833954.1 |
Target Gene Details
Entrez Gene ID: | 51025 |
Gene Name: | presequence translocase-associated motor 16 homolog (S. cerevisiae) |
Gene Aliases: |
CGI-136, MAGMAS, SMDMDM, TIM16, TIMM16 |
Location: |
Chr.16:4340251-4351372 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PAM16 | NM_016069.9 | NP_057153.8 | ||
| AF151894.1 | AAD34131.1 | |||
| AF349455.1 | AAL57767.1 | |||
| AK026514.1 | BAB15494.1 | |||
| BC005024.1 | AAH05024.1 | |||
| BG715429.1 | ||||
| CR457362.1 | CAG33643.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv471075 | Chr.16:4317729 - 4395326 on Build GRCh38 | Loss |
|
| nsv1046986 | Chr.16:4049000 - 5555721 on Build GRCh38 | Gain |
|
| nsv527866 | Chr.16:4206462 - 4412618 on Build GRCh38 | Loss |
|
| nsv571274 | Chr.16:4309420 - 4441189 on Build GRCh38 | Gain |
|
| esv3584924 | Chr.16:4330628 - 4469258 on Build GRCh38 | Gain |
|
| esv23732 | Chr.16:4340813 - 4476244 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72766573] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map