Assay Details
Target Gene Details
Entrez Gene ID: | 774 |
Gene Name: | calcium voltage-gated channel subunit alpha1 B |
Gene Aliases: |
BIII, CACNL1A5, CACNN, Cav2.2, DYT23 |
Location: |
Chr.9:137877789-138124624 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CACNA1B | NM_000718.3 | NP_000709.1 | ||
| NM_001243812.1 | NP_001230741.1 | |||
| XM_011518991.2 | XP_011517293.1 | |||
| XM_011518992.2 | XP_011517294.1 | |||
| XM_011518993.2 | XP_011517295.1 | |||
| XM_011518994.2 | XP_011517296.1 | |||
| XM_017015115.1 | XP_016870604.1 | |||
| XM_017015116.1 | XP_016870605.1 | |||
| XM_017015117.1 | XP_016870606.1 | |||
| XM_017015118.1 | XP_016870607.1 | |||
| XM_017015119.1 | XP_016870608.1 | |||
| XM_017015120.1 | XP_016870609.1 | |||
| M94172.1 | AAA51897.1 | |||
| M94173.1 | AAA51898.1 |
Target Gene Details
Entrez Gene ID: | 100133077 |
Gene Name: | uncharacterized LOC100133077 |
Gene Aliases: |
- |
Location: |
Chr.9:137867925-137892570 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100133077 | NR_121583.1 | |||
| AK128414.1 | BAC87428.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv145e19 | Chr.9:137878080 - 137883008 on Build GRCh38 | Loss |
|
| nsv415556 | Chr.9:137879161 - 137882744 on Build GRCh38 | Deletion |
|
| dgv4185n106 | Chr.9:137882748 - 137891148 on Build GRCh38 | Duplication |
|
| nsv1147231 | Chr.9:137882747 - 137891148 on Build GRCh38 | Duplication |
|
| nsv1076231 | Chr.9:137878110 - 137882736 on Build GRCh38 | Deletion |
|
| esv2666889 | Chr.9:137879158 - 137882742 on Build GRCh38 | Deletion |
|
| dgv4545e59 | Chr.9:137882679 - 137891477 on Build GRCh38 | Duplication |
|
| esv3622046 | Chr.9:137879214 - 137882785 on Build GRCh38 | Loss |
|
| nsv509338 | Chr.9:137839021 - 137984377 on Build GRCh38 | Insertion |
|
| nsv512132 | Chr.9:137879110 - 137884997 on Build GRCh38 | Loss |
|
| dgv48e194 | Chr.9:137878135 - 137882940 on Build GRCh38 | Deletion |
|
| dgv4184n106 | Chr.9:137878278 - 137882759 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map