Assay Details
Target Gene Details
Entrez Gene ID: | 5610 |
Gene Name: | eukaryotic translation initiation factor 2 alpha kinase 2 |
Gene Aliases: |
EIF2AK1, PKR, PPP1R83, PRKR |
Location: |
Chr.2:37099210-37157065 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF2AK2 | NM_001135651.2 | 12 | 1299 | NP_001129123.1 |
| NM_001135652.2 | 9 | 869 | NP_001129124.1 | |
| NM_002759.3 | 12 | 1533 | NP_002750.1 | |
| XM_011532987.2 | 12 | 1233 | XP_011531289.1 | |
| XM_017004503.1 | 12 | 1229 | XP_016859992.1 | |
| AK290655.1 | 12 | 1219 | ||
| AK313818.1 | 11 | 1056 | ||
| AY302136.1 | 9 | 853 | AAP57628.1 | |
| BC007769.2 | 11 | 1075 | AAH07769.1 | |
| BC022314.1 | ||||
| BC040851.1 | 12 | 1299 | AAH40851.1 | |
| BC057805.1 | 12 | 1204 | AAH57805.1 | |
| BC093676.1 | 10 | 1004 | AAH93676.1 | |
| BC101475.1 | 10 | 1004 | AAI01476.1 | |
| BC143230.1 | 10 | 1004 | ||
| M35663.1 | 11 | 1162 | AAA36409.1 | |
| M85294.1 | 10 | 1006 | AAA18253.1 |
More Information
Set Membership: |
|

Genomic Map