Assay Details
Target Gene Details
Entrez Gene ID: | 4926 |
Gene Name: | nuclear mitotic apparatus protein 1 |
Gene Aliases: |
NMP-22, NUMA |
Location: |
Chr.11:72002864-72080693 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUMA1 | NM_001286561.1 | 16 | 4001 | NP_001273490.1 |
| NM_006185.3 | 15 | 3903 | NP_006176.2 | |
| NR_104476.1 | ||||
| XM_006718564.1 | 14 | 3718 | XP_006718627.1 | |
| XM_011545054.1 | 14 | 3718 | XP_011543356.1 | |
| XM_011545055.1 | 15 | 3896 | XP_011543357.1 | |
| XM_011545056.2 | 16 | 3890 | XP_011543358.1 | |
| XM_011545057.2 | 16 | 3880 | XP_011543359.1 | |
| XM_011545058.1 | 14 | 3917 | XP_011543360.1 | |
| XM_011545059.2 | 16 | 3882 | XP_011543361.1 | |
| XM_011545060.2 | 16 | 3876 | XP_011543362.1 | |
| XM_011545061.2 | 14 | 3736 | XP_011543363.1 | |
| XM_011545062.2 | 14 | 3749 | XP_011543364.1 | |
| XM_011545063.2 | 14 | 3716 | XP_011543365.1 | |
| XM_011545064.1 | 14 | 3703 | XP_011543366.1 | |
| XM_011545065.1 | 14 | 3718 | XP_011543367.1 | |
| XM_011545066.1 | 14 | 3718 | XP_011543368.1 | |
| XM_017017830.1 | 15 | 3758 | XP_016873319.1 | |
| XM_017017831.1 | 14 | 3754 | XP_016873320.1 | |
| AB208841.1 | 1 | 2018 | BAD92078.1 | |
| AB210007.1 | 15 | 3704 | ||
| BC004165.2 | AAH04165.1 | |||
| BC008345.1 | 1 | 99 | AAH08345.1 | |
| BC013023.2 | 1 | 847 | AAH13023.1 | |
| Z11583.1 | 15 | 3721 | CAA77669.1 | |
| Z11584.1 | 16 | 3823 | CAA77670.1 | |
| Z14227.1 | 1 | 1738 | ||
| Z14228.1 | 1 | 643 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428262 | Chr.11:71652593 - 72093009 on Build GRCh38 | Gain+Loss |
|
| nsv951037 | Chr.11:72000055 - 72016354 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map