Assay Details
Target Gene Details
Entrez Gene ID: | 25906 |
Gene Name: | anaphase promoting complex subunit 15 |
Gene Aliases: |
APC15, C11orf51, HSPC020 |
Location: |
Chr.11:72106254-72112793 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Target Gene Details
Entrez Gene ID: | 220074 |
Gene Name: | leucine rich transmembrane and O-methyltransferase domain containing |
Gene Aliases: |
CFAP111, DFNB63, LRRC51 |
Location: |
Chr.11:72079982-72110782 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRTOMT | NM_001145308.4 | 7 | 3426 | NP_001138780.1 |
| NM_001145309.3 | 9 | 3647 | NP_001138781.1 | |
| NM_001145310.3 | 9 | 3527 | NP_001138782.1 | |
| XM_017017358.1 | 4 | 3481 | XP_016872847.1 | |
| AF289588.1 | 5 | 2508 | AAL55772.1 | |
| BX115135.1 | 2 | 452 | ||
| EU627069.1 | 9 | 3119 | ||
| EU627070.1 | 9 | 2999 |
More Information
Set Membership: |
|

Genomic Map